RIE156317826 Rieke Metals, Inc.

2-chloro-1-decene

  • MDL number MFCD00671868
  • Cas Number : 89632-03-1
  • Molecular Weight : 174.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/10004-002g

Safety Information

MSDS http://riekemetals.com/products-services/catnum/10004-002g
Availability details for
Quantity
  • Availability
  • Add to Cart