RIE156317835 Rieke Metals, Inc.

2-Bromo-1-heptene

  • MDL number MFCD00671859
  • Cas Number : 3252-91-3
  • Molecular Weight : 177.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/10004-003d

Safety Information

MSDS http://riekemetals.com/products-services/catnum/10004-003d
Availability details for
Quantity
  • Availability
  • Add to Cart