RIE156317895 Rieke Metals, Inc.

Ethyl 4-chlorobenzoylformate

  • MDL number MFCD01319615
  • Cas Number : 34966-48-8
  • Molecular Weight : 212.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/14009

Safety Information

MSDS http://riekemetals.com/products-services/catnum/14009
Availability details for
Quantity
  • Availability
  • Add to Cart