RIE156317922 Rieke Metals, Inc.

Ethyl 4-methylbenzoylformate

  • MDL number MFCD01311169
  • Cas Number : 5524-56-1
  • Molecular Weight : 192.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/14029

Safety Information

MSDS http://riekemetals.com/products-services/catnum/14029
Availability details for
Quantity
  • Availability
  • Add to Cart