RIE156317925 Rieke Metals, Inc.

Ethyl 2-naphthoyl formate

  • MDL number MFCD01934866
  • Cas Number : 73790-09-7
  • Molecular Weight : 228.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/14034

Safety Information

MSDS http://riekemetals.com/products-services/catnum/14034
Availability details for
Quantity
  • Availability
  • Add to Cart