RIE156317927 Rieke Metals, Inc.

Ethyl 4-bromobenzoyl formate

  • MDL number MFCD06659764
  • Cas Number : 20201-26-7
  • Molecular Weight : 257.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/14036

Safety Information

MSDS http://riekemetals.com/products-services/catnum/14036
Availability details for
Quantity
  • Availability
  • Add to Cart