RIE156317957 Rieke Metals, Inc.

Ethyl 3,4-dichlorobenzoylformate

  • MDL number MFCD01934864
  • Cas Number : 34966-52-4
  • Molecular Weight : 247.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/14066

Safety Information

MSDS http://riekemetals.com/products-services/catnum/14066
Availability details for
Quantity
  • Availability
  • Add to Cart