RIE156317983 Rieke Metals, Inc.

2-Bromo-3-(3,5-difluorophenyl)-1-propene

  • MDL number MFCD06201206
  • Cas Number : 842140-35-6
  • Molecular Weight : 233.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/10005-743a

Safety Information

MSDS http://riekemetals.com/products-services/catnum/10005-743a
Availability details for
Quantity
  • Availability
  • Add to Cart