RIE156318017 Rieke Metals, Inc.

2-bromo-3-(3-methylphenyl)-1-propene

  • MDL number MFCD00671931
  • Cas Number : 731772-18-2
  • Molecular Weight : 211.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/10005-083a

Safety Information

MSDS http://riekemetals.com/products-services/catnum/10005-083a
Availability details for
Quantity
  • Availability
  • Add to Cart