RIE156318773 Rieke Metals, Inc.

trans-2-(3-chlorobenzoyl)cyclohexane-1-carboxylic acid

  • MDL number MFCD01311270
  • Cas Number : 733742-78-4
  • Molecular Weight : 266.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/18510-20

Safety Information

MSDS http://riekemetals.com/products-services/catnum/18510-20
Availability details for
Quantity
  • Availability
  • Add to Cart