RIE156318982 Rieke Metals, Inc.

trans-4-[2-oxo-2-(3-nitrophenyl)ethyl]cyclohexane-1-carboxylic acid

  • MDL number MFCD06799215
  • Molecular Weight : 291.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/18520-47

Safety Information

MSDS http://riekemetals.com/products-services/catnum/18520-47
Availability details for
Quantity
  • Availability
  • Add to Cart