RIE156318984 Rieke Metals, Inc.

Methyl trans-2-hydroxymethylcyclohexane-1-carboxylate

  • MDL number MFCD06799218
  • Cas Number : 71550-80-6
  • Molecular Weight : 172.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/18522

Safety Information

MSDS http://riekemetals.com/products-services/catnum/18522
Availability details for
Quantity
  • Availability
  • Add to Cart