RIE156318986 Rieke Metals, Inc.

Methyl cis-4-hydroxymethylcyclohexane-1-carboxylate

  • MDL number MFCD06799220
  • Cas Number : 13380-85-3
  • Molecular Weight : 172.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 4-(hydroxymethyl)cyclohexanecarboxylate
Inchi Code 1S/C9H16O3/c1-12-9(11)8-4-2-7(6-10)3-5-8/h7-8,10H,2-6H2,1H3/t7-,8+
Inchi Key KOGYKIDJFOMAOF-OCAPTIKFSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/18525

Safety Information

MSDS http://riekemetals.com/products-services/catnum/18525
Availability details for
Quantity
  • Availability
  • Add to Cart