RIE156318987 Rieke Metals, Inc.

Ethyl trans-4-(N-benzoylaminomethyl)cyclohexane-1-carboxylic acid

  • MDL number MFCD07698707
  • Molecular Weight : 289.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/18532

Safety Information

MSDS http://riekemetals.com/products-services/catnum/18532
Availability details for
Quantity
  • Availability
  • Add to Cart