RIE156318992 Rieke Metals, Inc.

4-Chloro-3-fluorophenylzinc iodide

  • MDL number MFCD01319863
  • Cas Number : 312693-42-8
  • Molecular Weight : 321.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2047

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2047
Availability details for
Quantity
  • Availability
  • Add to Cart