RIE156318998 Rieke Metals, Inc.

1-Hexadecylzinc bromide

  • MDL number MFCD02260127
  • Cas Number : 737797-10-3
  • Molecular Weight : 370.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2142

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2142
Availability details for
Quantity
  • Availability
  • Add to Cart