RIE156319002 Rieke Metals, Inc.

2,5-Dichlorobenzylzinc chloride

  • MDL number MFCD02260137
  • Cas Number : 737797-20-5
  • Molecular Weight : 260.85

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2152

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2152
Availability details for
Quantity
  • Availability
  • Add to Cart