RIE156319012 Rieke Metals, Inc.

3,4-Difluorophenylzinc bromide

  • MDL number MFCD02260154
  • Cas Number : 737797-28-3
  • Molecular Weight : 258.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2167

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2167
Availability details for
Quantity
  • Availability
  • Add to Cart