RIE156319016 Rieke Metals, Inc.

3-Bromo-2-thienylzinc bromide

  • MDL number MFCD02260168
  • Cas Number : 672325-70-1
  • Molecular Weight : 307.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2181

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2181
Availability details for
Quantity
  • Availability
  • Add to Cart