RIE156319017 Rieke Metals, Inc.

5-Chloro-2-methoxyphenylzinc iodide

  • MDL number MFCD02260171
  • Cas Number : 737797-36-3
  • Molecular Weight : 333.85

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2184

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2184
Availability details for
Quantity
  • Availability
  • Add to Cart