RIE156319020 Rieke Metals, Inc.

2,4,6-Trimethylphenylzinc iodide

  • MDL number MFCD07698711
  • Cas Number : 718628-26-3
  • Molecular Weight : 311.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2194

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2194
Availability details for
Quantity
  • Availability
  • Add to Cart