RIE156319021 Rieke Metals, Inc.

4-tert-Butylbenzylzinc bromide

  • MDL number MFCD02260184
  • Cas Number : 737797-48-7
  • Molecular Weight : 292.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2199

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2199
Availability details for
Quantity
  • Availability
  • Add to Cart