RIE156319022 Rieke Metals, Inc.

4-Trifluoromethylbenzylzinc chloride

  • MDL number MFCD07698712
  • Cas Number : 738580-34-2
  • Molecular Weight : 259.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2201

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2201
Availability details for
Quantity
  • Availability
  • Add to Cart