RIE156319028 Rieke Metals, Inc.

2-[(4-Morpholino)methyl]phenylzinc iodide

  • MDL number MFCD07698718
  • Molecular Weight : 368.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2215

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2215
Availability details for
Quantity
  • Availability
  • Add to Cart