RIE156319029 Rieke Metals, Inc.

3-[(4-Morpholino)methyl]phenylzinc iodide

  • MDL number MFCD07698719
  • Molecular Weight : 368.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2216

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2216
Availability details for
Quantity
  • Availability
  • Add to Cart