RIE156319032 Rieke Metals, Inc.

3-[(1-Pyrrolidino)methyl]phenylzinc iodide

  • MDL number MFCD07698722
  • Molecular Weight : 352.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2219

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2219
Availability details for
Quantity
  • Availability
  • Add to Cart