RIE156319034 Rieke Metals, Inc.

2-[(1-Piperidino)methyl]phenylzinc iodide

  • MDL number MFCD07698724
  • Molecular Weight : 366.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2221

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2221
Availability details for
Quantity
  • Availability
  • Add to Cart