RIE156319037 Rieke Metals, Inc.

4-Chlorophenethylzinc bromide

  • MDL number MFCD07698728
  • Molecular Weight : 284.88

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2225

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2225
Availability details for
Quantity
  • Availability
  • Add to Cart