RIE156319042 Rieke Metals, Inc.

3-Methylphenethylzinc bromide

  • MDL number MFCD07698733
  • Molecular Weight : 264.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2230

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2230
Availability details for
Quantity
  • Availability
  • Add to Cart