RIE156319046 Rieke Metals, Inc.

2-Methoxyphenethylzinc bromide

  • MDL number MFCD07698737
  • Molecular Weight : 280.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2234

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2234
Availability details for
Quantity
  • Availability
  • Add to Cart