RIE156319047 Rieke Metals, Inc.

Neopentylzinc iodide

  • MDL number MFCD07698738
  • Cas Number : 262422-94-6
  • Molecular Weight : 263.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2235

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2235
Availability details for
Quantity
  • Availability
  • Add to Cart