RIE156319048 Rieke Metals, Inc.

n-Dodecylzinc bromide

  • MDL number MFCD07698740
  • Cas Number : 869589-06-0
  • Molecular Weight : 314.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2237

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2237
Availability details for
Quantity
  • Availability
  • Add to Cart