RIE156319049 Rieke Metals, Inc.

3,5-Dichlorobenzylzinc chloride

  • MDL number MFCD02260138
  • Cas Number : 352530-34-8
  • Molecular Weight : 260.85

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2238

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2238
Availability details for
Quantity
  • Availability
  • Add to Cart