RIE156319055 Rieke Metals, Inc.

3-Phenyl-1-propylzinc bromide

  • MDL number MFCD08064069
  • Cas Number : 626207-42-9
  • Molecular Weight : 264.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2245

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2245
Availability details for
Quantity
  • Availability
  • Add to Cart