RIE156319057 Rieke Metals, Inc.

4-Bromo-3-methylphenylzinc iodide

  • MDL number MFCD08064071
  • Molecular Weight : 362.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2247

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2247
Availability details for
Quantity
  • Availability
  • Add to Cart