RIE156319062 Rieke Metals, Inc.

7-Ethoxy-7-oxoheptylzinc bromide

  • MDL number MFCD08741501
  • Molecular Weight : 302.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2255

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2255
Availability details for
Quantity
  • Availability
  • Add to Cart