RIE156319071 Rieke Metals, Inc.

4-n-Butylphenylmagnesium bromide

  • MDL number MFCD01317004
  • Cas Number : 185416-14-2
  • Molecular Weight : 237.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/3014

Safety Information

MSDS http://riekemetals.com/products-services/catnum/3014
Availability details for
Quantity
  • Availability
  • Add to Cart