RIE156319072 Rieke Metals, Inc.

6-Methyl-2-pyridylmagnesium bromide

  • MDL number MFCD01311478
  • Cas Number : 621685-64-1
  • Molecular Weight : 196.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/3028

Safety Information

MSDS http://riekemetals.com/products-services/catnum/3028
Availability details for
Quantity
  • Availability
  • Add to Cart