RIE156319073 Rieke Metals, Inc.

2,3,5,6-Tetramethylphenylmagnesium bromide

  • MDL number MFCD01311498
  • Cas Number : 75724-98-0
  • Molecular Weight : 237.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/3047

Safety Information

MSDS http://riekemetals.com/products-services/catnum/3047
Availability details for
Quantity
  • Availability
  • Add to Cart