RIE156319082 Rieke Metals, Inc.

4-Phenylbenzylmagnesium chloride

  • MDL number MFCD02260203
  • Cas Number : 130387-76-7
  • Molecular Weight : 227.00

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/3106

Safety Information

MSDS http://riekemetals.com/products-services/catnum/3106
Availability details for
Quantity
  • Availability
  • Add to Cart