RIE156319084 Rieke Metals, Inc.

2-Ethoxybenzylmagnesium chloride

  • MDL number MFCD02260217
  • Cas Number : 738580-60-4
  • Molecular Weight : 194.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/3120

Safety Information

MSDS http://riekemetals.com/products-services/catnum/3120
Availability details for
Quantity
  • Availability
  • Add to Cart