RIE156319086 Rieke Metals, Inc.

2-Ethylphenylmagnesium bromide

  • MDL number MFCD02260221
  • Cas Number : 21450-63-5
  • Molecular Weight : 209.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/3124

Safety Information

MSDS http://riekemetals.com/products-services/catnum/3124
Availability details for
Quantity
  • Availability
  • Add to Cart