RIE156319088 Rieke Metals, Inc.

3-Quinolylmagnesium bromide

  • MDL number MFCD02260230
  • Cas Number : 738580-61-5
  • Molecular Weight : 232.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/3134

Safety Information

MSDS http://riekemetals.com/products-services/catnum/3134
Availability details for
Quantity
  • Availability
  • Add to Cart