RIE156319089 Rieke Metals, Inc.

3-Bromo-2-thienylmagnesium bromide

  • MDL number MFCD02260236
  • Cas Number : 119277-44-0
  • Molecular Weight : 266.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/3141

Safety Information

MSDS http://riekemetals.com/products-services/catnum/3141
Availability details for
Quantity
  • Availability
  • Add to Cart