RIE156319095 Rieke Metals, Inc.

3-(1,3-Dioxan-2-yl)-4-fluorophenylmagnesium bromide

  • MDL number MFCD03840900
  • Molecular Weight : 285.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/3160

Safety Information

MSDS http://riekemetals.com/products-services/catnum/3160
Availability details for
Quantity
  • Availability
  • Add to Cart