RIE156319099 Rieke Metals, Inc.

2-[8-(1,4-Dioxa-8-azaspiro[4.5]decyl)methyl]phenylmagnesium bromide

  • MDL number MFCD06200645
  • Molecular Weight : 336.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/3167

Safety Information

MSDS http://riekemetals.com/products-services/catnum/3167
Availability details for
Quantity
  • Availability
  • Add to Cart