RIE156319104 Rieke Metals, Inc.

4-Fluoro-3-methylbenzylmagnesium chloride

  • MDL number MFCD07698752
  • Molecular Weight : 182.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/3173

Safety Information

MSDS http://riekemetals.com/products-services/catnum/3173
Availability details for
Quantity
  • Availability
  • Add to Cart