RIE156319106 Rieke Metals, Inc.

4-Chloro-2-methylbenzylmagnesium chloride

  • MDL number MFCD08064074
  • Molecular Weight : 199.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/3175

Safety Information

MSDS http://riekemetals.com/products-services/catnum/3175
Availability details for
Quantity
  • Availability
  • Add to Cart