RIE156319108 Rieke Metals, Inc.

3-Fluoro-4-methoxybenzylmagnesium chloride

  • MDL number MFCD07698754
  • Molecular Weight : 198.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/3177

Safety Information

MSDS http://riekemetals.com/products-services/catnum/3177
Availability details for
Quantity
  • Availability
  • Add to Cart