RIE156319113 Rieke Metals, Inc.

2,4,5-Trimethylbenzylmagnesium chloride

  • MDL number MFCD07698756
  • Molecular Weight : 192.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/3183

Safety Information

MSDS http://riekemetals.com/products-services/catnum/3183
Availability details for
Quantity
  • Availability
  • Add to Cart